Potassium phosphate, tribasic monohydrate, 96%, extra pure
Potassium phosphate, tribasic monohydrate, 96%, extra pure
Potassium phosphate, tribasic monohydrate, 96%, extra pure
Potassium phosphate, tribasic monohydrate, 96%, extra pure
Thermo Scientific Chemicals
Potassium phosphate, tribasic monohydrate, 96%, extra pure
CAS: 27176-10-9 | K3O4P·H2O | 230.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA263795000
Price (MYR)
366.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
366.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium phosphate, tribasic monohydrate
Melting Point1340.0°C
Name Notec.p.
CAS27176-10-9