Zinc stearate
Zinc stearate
Zinc stearate
Thermo Scientific Chemicals
Zinc stearate
CAS: 557-05-1 | C36H70O4Zn | 632.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA263780010
Price (MYR)
321.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
321.00
BOT
Add to cart
Specifications
Chemical Name or MaterialZinc stearate
CAS557-05-1
Melting Point118.0°C to 128.0°C
ColorWhite
RTECS NumberZH5200000