L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
L(-)-Camphorsulfonic acid, 98%
Thermo Scientific Chemicals
L(-)-Camphorsulfonic acid, 98%
CAS: 35963-20-3 | C10H16O4S | 232.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA263641000
Price (MYR)
873.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
873.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(-)-Camphorsulfonic acid
CAS35963-20-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre