Tetrahydro-2-furoic acid, 98+%
Tetrahydro-2-furoic acid, 98+%
Tetrahydro-2-furoic acid, 98+%
Tetrahydro-2-furoic acid, 98+%
Thermo Scientific Chemicals
Tetrahydro-2-furoic acid, 98+%
CAS: 16874-33-2 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA263141000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrahydro-2-furoic acid
Name Note99+%
CAS16874-33-2
CAS Min %99.0
ColorYellow to Brown