Hydroxychloroquine sulfate, 98%
Hydroxychloroquine sulfate, 98%
Hydroxychloroquine sulfate, 98%
Hydroxychloroquine sulfate, 98%
Thermo Scientific Chemicals
Hydroxychloroquine sulfate, 98%
Hydroxychloroquine sulfate, CAS # 747-36-4, is a chemical compound with antiparasitic, antiviral, and anti-rheumatic activities. | CAS: 747-36-4 | C18H26ClN3O·H2SO4 | 433.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA263011000
Price (MYR)
3,180.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,180.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydroxychloroquine sulfate
CAS747-36-4
Health Hazard 1Serious eye damage/eye irritation (category 2), Acute toxicity (category 4)
Health Hazard 2Causes serious eye irritation, Harmful if swallowed
Melting Point240.0°C
View more
General Description
  • Hydroxychloroquine sulfate shows antimalarial activity against malaria parasites mainly in the intra-erythrocytic phase of its life cycle. It acts by inhibiting the heme polymerase inside the food vacuole of the parasites
  • It shows an inhibitory activity in autophagy preventing the lysosome fusion with the autophagosome
  • Some studies demonstrated that hydroxychloroquine sulfate, due to its antiviral activity can restrict HIV infection in vitro
Applications
  • Hydroxychloroquine sulfate displays antiviral and anti-rheumatic activities
  • In vitro and in vivo experiments have been performed to investigate the role of hydroxychloroquine sulfate in the treatment of SARS-CoV-2 disease
  • Hydroxychloroquine sulfate can be used in immunofluorescence assays to evaluate the antiviral mechanisms in dengue virus-2 infection. In this disease, this compound can activate the innate immune response
RUO – Research Use Only