L(+)-Selenomethionine, 99+%, ee 99+%
L(+)-Selenomethionine, 99+%, ee 99+%
L(+)-Selenomethionine, 99+%, ee 99+%
L(+)-Selenomethionine, 99+%, ee 99+%
Thermo Scientific Chemicals
L(+)-Selenomethionine, 99+%, ee 99+%
CAS: 3211-76-5 | C5H11NO2Se | 196.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA259962500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialL(+)-Selenomethionine
Name Note99+% e.e.
CAS3211-76-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.