3,7-Dimethyl-7-hydroxyoctanal, 97%
3,7-Dimethyl-7-hydroxyoctanal, 97%
3,7-Dimethyl-7-hydroxyoctanal, 97%
3,7-Dimethyl-7-hydroxyoctanal, 97%
Thermo Scientific Chemicals
3,7-Dimethyl-7-hydroxyoctanal, 97%
CAS: 107-75-5 | C10H20O2 | 172.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA25968025025 mLGlass bottle
Catalog number FSA259680250
Price (MYR)
356.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
356.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 7-Dimethyl-7-hydroxyoctanal
Name Note98%
CAS107-75-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger