Potassium tetraborate tetrahydrate, 99.5%, extra pure
Potassium tetraborate tetrahydrate, 99.5%, extra pure
Potassium tetraborate tetrahydrate, 99.5%, extra pure
Potassium tetraborate tetrahydrate, 99.5%, extra pure
Thermo Scientific Chemicals
Potassium tetraborate tetrahydrate, 99.5%, extra pure
CAS: 12045-78-2 | B4K2O7·4H2O | 305.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA259570010
Price (MYR)
498.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
498.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPotassium tetraborate tetrahydrate
Melting Point850.0°C
CAS12045-78-2
ColorWhite
Health Hazard 1GHS Signal Word: Warning