1-Ethoxy-2-propanol, 95%
1-Ethoxy-2-propanol, 95%
1-Ethoxy-2-propanol, 95%
1-Ethoxy-2-propanol, 95%
Thermo Scientific Chemicals
1-Ethoxy-2-propanol, 95%
CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA259050025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Ethoxy-2-propanol
CAS1569-02-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.