Gum arabic, powder
Thermo Scientific Chemicals
Gum arabic, powder
CAS: 9000-01-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA258852500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGum arabic
ColorBeige-Yellow to White
CAS9000-01-5
Physical FormPowder
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.