Theobromine, 99%
Theobromine, 99%
Theobromine, 99%
Thermo Scientific Chemicals
Theobromine, 99%
Theobromine, CAS # 83-67-0, is a methylxanthine alkaloid that is present in large quantities in cocoa and is a derivative of caffein. | CAS: 83-67-0 | C7H8N4O2 | 180.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic bottle
Catalog number FSA258822500
Price (MYR)
970.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
970.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTheobromine
Name Note99%
CAS83-67-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
View more
General description
• Theobromine is a methylxanthine alkaloid that is a derivative of caffeine
Applications
• Theobromine is a phosphodiesterase inhibitor, weak adenosine receptor antagonist, and smooth muscle relaxant
• As a catabolic product of caffeine, it has antioxidant and prooxidant properties
• It can be used as a standard for quantification of theobromine
RUO – Research Use Only
General References:
  1. Johannsen, M.; Brunner, G. Solubilities of the xanthines caffeine, theophylline and theobromine in supercritical carbon dioxide. Fluid Phase Equilibria.,1994, 95(1), 215-226.
  2. Kreiser, W.R.; Martin, R.A. Jr. High pressure liquid chromatographic determination of theobromine and caffeine in cocoa and chocolate products. J Assoc Off Anal Chem. 1978, 61(6), 1424-1427.