(Aminomethyl)cyclopropane, 97%
(Aminomethyl)cyclopropane, 97%
(Aminomethyl)cyclopropane, 97%
(Aminomethyl)cyclopropane, 97%
Thermo Scientific Chemicals
(Aminomethyl)cyclopropane, 97%
CAS: 2516-47-4 | C4H9N | 71.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA258720050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(Aminomethyl)cyclopropane
CAS2516-47-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapour.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi