Buffer solution pH 5, Acetate buffer
Buffer solution pH 5, Acetate buffer
Buffer solution pH 5, Acetate buffer
Buffer solution pH 5, Acetate buffer
Thermo Scientific Chemicals
Buffer solution pH 5, Acetate buffer
CAS: 64-19-7
Have Questions?
Catalog NumberQuantity
FSA2586200101 L
Catalog number FSA258620010
Price (MYR)
338.00
BOT
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
338.00
BOT
Add to cart
Chemical Identifiers
CAS64-19-7, 7732-18-5, 1310-58-3
Specifications
Chemical Name or MaterialBuffer solution pH 5
CAS64-19-7
ColorColorless
CAS7732-18-5
Density1.0000g/mL
View more
pH buffer
RUO – Research Use Only