Ibuprofen, 99%, Thermo Scientific Chemicals
Ibuprofen, 99%, Thermo Scientific Chemicals
Ibuprofen, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ibuprofen, 99%, Thermo Scientific Chemicals
Ibuprofen, 2-4-isobutylphenyl propanoic acid, CAS # 15687-27-1, is a laboratory form of the over-the-counter nonsteroidal anti-inflammatory (NSAID) drug.
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
50 g
Packaging:
Glass bottle
Catalog number FSA258610100
Price (MYR)
1,428.00
Bottle
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,428.00
Bottle
Specifications
Chemical Name or MaterialIbuprofen
CAS15687-27-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
View more
General description
  • Ibuprofen (2-4-isobutylphenyl propanoic acid) is a common NSAID that reduces pyrogen release and inflammation
  • Ibuprofen is a racemic mixture that consists of an R-enantiomer and S-enantiomer; the S-enantiomer is proposed to be the stronger acting agent
  • It is a propionic acid derivative, a short-chain saturated fatty acid bound to ethane
  • Ibuprofen inhibits cyclooxygenases and reduces prostaglandin formation, inhibiting fever and pain
Applications
  • Ibuprofen reduces both local and systemic inflammation; suitable for use in vitro or in vivo to study specific or broad mechanisms
  • Intended for sensitive biochemical applications such as HPLC
RUO – Research Use Only