Buffer solution pH 7, Phosphate buffer
Buffer solution pH 7, Phosphate buffer
Buffer solution pH 7, Phosphate buffer
Buffer solution pH 7, Phosphate buffer
Thermo Scientific Chemicals
Buffer solution pH 7, Phosphate buffer
CAS: 7558-79-4 | H2KO4P
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
2.5 L
Catalog number FSA258595000
Price (MYR)
331.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
331.00
BOT
Add to cart
Chemical Identifiers
CAS7558-79-4, 7732-18-5
Specifications
Chemical Name or MaterialBuffer solution pH 7
CAS7558-79-4
CAS7732-18-5
Density1.0030g/mL
Quantity500 mL
View more
pH buffer
RUO – Research Use Only