1-Octacosanol, 90+%, Thermo Scientific Chemicals
1-Octacosanol, 90+%, Thermo Scientific Chemicals
1-Octacosanol, 90+%, Thermo Scientific Chemicals
1-Octacosanol, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Octacosanol, 90+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
Packaging:
Glass bottle
Catalog number FSA258132500
Price (MYR)
1,149.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,149.00
EA
Add to cart
Specifications
Chemical Name or Material1-Octacosanol
CAS557-61-9
CAS Min %90.0
CAS Max %100.0
Melting Point83.0°C