Oxytetracycline, 'can Be Used as Secondarystandard'
Oxytetracycline, 'can Be Used as Secondarystandard'
Oxytetracycline, 'can Be Used as Secondarystandard'
Oxytetracycline, 'can Be Used as Secondarystandard'
Thermo Scientific Chemicals
Oxytetracycline, 'can Be Used as Secondarystandard'
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
Packaging:
Glass Bottle
Catalog number FSA257705000
Price (MYR)
926.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
926.00
BOT
Add to cart
Specifications
Chemical Name or MaterialOxytetracycline
TypeOxytetracycline
Physical FormCrystalline Powder
CAS79-57-2
Melting Point183.0°C