Sodium tert-butoxide, 99%, pure
Sodium tert-butoxide, 99%, pure
Sodium tert-butoxide, 99%, pure
Sodium tert-butoxide, 99%, pure
Thermo Scientific Chemicals
Sodium tert-butoxide, 99%, pure
CAS: 865-48-5 | C4H9NaO | 96.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA252555000
Price (MYR)
1,397.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,397.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium tert-butoxide
CAS865-48-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Self-heating; may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.
Flammable solid.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f