4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
Thermo Scientific Chemicals
4-Amino-6-chloro-1,3-benzenedisulfonamide, 98%
CAS: 121-30-2 | C6H8ClN3O4S2 | 285.73 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA251850050
Price (MYR)
264.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
264.00
EA
Add to cart
Specifications
Chemical Name or Material4-Amino-6-chloro-1, 3-benzenedisulfonamide
CAS121-30-2
CAS Min %97.5
CAS Max %100.0
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.