Citric acid, disodium salt sesquihydrate, 99%, pure
Citric acid, disodium salt sesquihydrate, 99%, pure
Citric acid, disodium salt sesquihydrate, 99%, pure
Citric acid, disodium salt sesquihydrate, 99%, pure
Thermo Scientific Chemicals
Citric acid, disodium salt sesquihydrate, 99%, pure
CAS: 1545832 | C12H18Na4O17 | 526.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA250242500
Price (MYR)
296.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
296.00
EA
Add to cart
Specifications
Chemical Name or MaterialCitric acid, disodium salt sesquihydrate
Name NotePure
CAS1545832
Melting Point149.0°C