1-Iodododecane, 98%, stabilized
1-Iodododecane, 98%, stabilized
1-Iodododecane, 98%, stabilized
1-Iodododecane, 98%, stabilized
Thermo Scientific Chemicals
1-Iodododecane, 98%, stabilized
CAS: 4292-19-7 | C12H25I | 296.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA250091000
Price (MYR)
1,055.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,055.00
EA
Add to cart
Specifications
Chemical Name or Material1-Iodododecane
Name NoteStabilized with 500ppm copper
CAS4292-19-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning