Ethyl potassium malonate, 98%
Ethyl potassium malonate, 98%
Ethyl potassium malonate, 98%
Ethyl potassium malonate, 98%
Thermo Scientific Chemicals
Ethyl potassium malonate, 98%
CAS: 6148-64-7 | C5H7KO4 | 170.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA248080250
Price (MYR)
287.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
287.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl potassium malonate
Name Note98%
CAS6148-64-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: