2-Amino-5-nitrobenzoic acid, 98%
2-Amino-5-nitrobenzoic acid, 98%
2-Amino-5-nitrobenzoic acid, 98%
2-Amino-5-nitrobenzoic acid, 98%
Thermo Scientific Chemicals
2-Amino-5-nitrobenzoic acid, 98%
CAS: 616-79-5 | C7H6N2O4 | 182.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA245950050
Price (MYR)
304.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
304.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Amino-5-nitrobenzoic acid
Name Note98%
CAS616-79-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning