1-Methoxy-2-propanol, 98.5%, extra pure
1-Methoxy-2-propanol, 98.5%, extra pure
1-Methoxy-2-propanol, 98.5%, extra pure
1-Methoxy-2-propanol, 98.5%, extra pure
Thermo Scientific Chemicals
1-Methoxy-2-propanol, 98.5%, extra pure
CAS: 107-98-2 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
25 mL
1 L
2.5 L
5 L
Packaging:
View all
Metal drum
Catalog number FSA244990251
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Specifications
Chemical Name or Material1-Methoxy-2-propanol
CAS107-98-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest