Potassium citrate tribasic monohydrate, 99%, extra pure
Potassium citrate tribasic monohydrate, 99%, extra pure
Potassium citrate tribasic monohydrate, 99%, extra pure
Potassium citrate tribasic monohydrate, 99%, extra pure
Thermo Scientific Chemicals
Potassium citrate tribasic monohydrate, 99%, extra pure
CAS: 6100-05-6 | C6H5K3O7·H2O | 324.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA244210010
Price (MYR)
475.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
475.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium citrate tribasic monohydrate
CAS6100-05-6
Melting Point180.0°C
Assay Percent Range99%
PackagingPlastic bottle