n-Hexane-d14, for NMR, 99+ atom % D
n-Hexane-d14, for NMR, 99+ atom % D
n-Hexane-d14, for NMR, 99+ atom % D
n-Hexane-d14, for NMR, 99+ atom % D
Thermo Scientific Chemicals
n-Hexane-d14, for NMR, 99+ atom % D
CAS: 21666-38-6 | C6H14 | 100.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass Bottle
Catalog number FSA244010010
Price (MYR)
764.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
764.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Hexane-d14
Density0.9600g/mL
Deuterium (D)99.50% min.
PackagingGlass Bottle
Physical FormLiquid
View more
For NMR
RUO – Research Use Only