Kjeldahl tablets with selenium (Wieninger)
Kjeldahl tablets with selenium (Wieninger)
Kjeldahl tablets with selenium (Wieninger)
Kjeldahl tablets with selenium (Wieninger)
Thermo Scientific Chemicals
Kjeldahl tablets with selenium (Wieninger)
CAS: 7782-49-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA243920010
Price (MYR)
1,806.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,806.00
BOT
Add to cart
Chemical Identifiers
CAS7782-49-2, 7758-99-8, 7757-82-6
Specifications
Chemical Name or MaterialKjeldahl tablets with selenium (Wieninger)
CAS7782-49-2
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.
CAS7758-99-8
Health Hazard 3GHS P Statement
Avoid release to the environment.