D-Calcium pantothenate, 98%
D-Calcium pantothenate, 98%
D-Calcium pantothenate, 98%
D-Calcium pantothenate, 98%
Thermo Scientific Chemicals
D-Calcium pantothenate, 98%
CAS: 137-08-6 | C18H32CaN2O10 | 476.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic Bottle
Catalog number FSA243305000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Calcium pantothenate
Melting Point190°C
CAS137-08-6
ColorWhite
EINECS Number205-278-9