Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.060-0.200 mm, 40 A
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA241660010
Price (MYR)
889.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
889.00
BOT
Add to cart
Specifications
Pore Size40 Å
PackagingPlastic Bottle
Quantity1 kg
TypeSilica Gel
MaterialSilica Gel
View more
For column chromatography
RUO – Research Use Only