Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Silica gel, for chromatography, 0.035-0.070 mm, 90 A
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.035-0.070 mm, 90 A
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Plastic Bottle
Catalog number FSA241652500
Price (MYR)
325.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
325.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilica gel
Name NoteFor chromatography, 0.035-0.070mm, 90Å
CAS7631-86-9
MaterialSilica Gel
Quantity250 g
View more
For column chromatography
RUO – Research Use Only