Sesame oil
Thermo Scientific Chemicals
Sesame oil
CAS: 8008-74-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Packaging:
Glass bottle
Catalog number FSA241000010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSesame oil
Acidity0.6mg KOH/g max.
CAS8008-74-0
Melting Point-5.0°C
ColorYellow