Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.060-0.200 mm, 60 A
For column chromatography. | CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 kg
5 kg
Catalog number FSA240370300
Price (MYR)
11,699.00
BOT
Add to cart
Quantity:
30 kg
Request bulk or custom format
Price (MYR)
11,699.00
BOT
Add to cart
Specifications
Quantity30 kg
TypeSilica Gel
MaterialSilica Gel
For Use With (Application)Column Chromatography
Molecular FormulaO2Si