Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.035-0.070 mm, 60 A
For column chromatography. | CAS: 7631-86-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number FSA240360050
Price (MYR)
3,367.00
BOT
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
3,367.00
BOT
Add to cart
Specifications
Pore Size60 Å
PackagingPlastic Drum
Quantity5 kg
TypeSilica Gel
MaterialSilica Gel
View more
For column chromatography
RUO – Research Use Only