Agarose, for biochemistry, pure powder (Mr = - 0.17)
CAS: 9012-36-6
Have Questions?
Change view
Quantity:
500 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number FSA239025000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAgarose
Name NotePure powder (Mr = - 0.17), For biochemistry
Physical FormPowder
CAS9012-36-6