L-Buthionine-(S,R)-sulfoximine, 99%
L-Buthionine-(S,R)-sulfoximine, 99%
L-Buthionine-(S,R)-sulfoximine, 99%
Thermo Scientific Chemicals
L-Buthionine-(S,R)-sulfoximine, 99%
CAS: 83730-53-4 | C8H18N2O3S | 222.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA235520050
Price (MYR)
5,489.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,489.00
Each
Specifications
Chemical Name or MaterialL-Buthionine-(S, R)-sulfoximine
CAS83730-53-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee