Inositol hexaphosphoric acid, 50 wt% aqueous solution
Inositol hexaphosphoric acid, 50 wt% aqueous solution
Inositol hexaphosphoric acid, 50 wt% aqueous solution
Inositol hexaphosphoric acid, 50 wt% aqueous solution
Thermo Scientific Chemicals
Inositol hexaphosphoric acid, 50 wt% aqueous solution
CAS: 83-86-3 | C6H18O24P6 | 660.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number FSA235372500
Price (MYR)
487.00
BOT
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
487.00
BOT
Add to cart
Chemical Identifiers
CAS83-86-3, 7732-18-5
Specifications
Chemical Name or MaterialInositol hexaphosphoric acid
Name Note40-50 wt. % Aqueous Solution
CAS83-86-3
Boiling Point105.0°C
Health Hazard 1Warning