Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
Thermo Scientific Chemicals
Glutaric dialdehyde, electron microscopy grade, 25% solution in water, purified
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA233281000
Price (MYR)
1,210.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,210.00
EA
Add to cart
Chemical Identifiers
CAS111-30-8, 7732-18-5
Specifications
Chemical Name or MaterialGlutaric dialdehyde, electron microscopy grade
Name NotePurified
CAS111-30-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Very t