4-Epitetracycline hydrochloride, can be used as a secondary standard
4-Epitetracycline hydrochloride, can be used as a secondary standard
4-Epitetracycline hydrochloride, can be used as a secondary standard
4-Epitetracycline hydrochloride, can be used as a secondary standard
Thermo Scientific Chemicals
4-Epitetracycline hydrochloride, can be used as a secondary standard
4-Epitetracycline hydrochloride, CAS # 23313-80-6, is a degradation product of tetracycline that shows antibiotic activity and can induce the protein biosynthesis inhibition.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA233121000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Epitetracycline hydrochloride
CAS23313-80-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Suspected of damaging the unborn child.
Health Hazard 3GHS P Statement:
Use personal protective equipment as required.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
General Description
  • 4-Epitetracycline hydrochloride is a metabolite derived from the degradation of tetracycline compounds and has been shown to display antibiotic activity
  • This compound blocks protein synthesis by binding to the 30S ribosomal subunit and interfering with the binding of aminoacyl-tRNA to the mRNA-ribosome complex
Applications
  • This semi-synthesized compound derived from actinomycetes can decrease the activity of the Tet repressor effector in animal models
  • In vivo experiments revealed that 4-epitetracycline hydrochloride can induce a toxic response
RUO – Research Use Only