Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Petroleum ether, extra pure, boiling range 100-140°C
Thermo Scientific Chemicals
Petroleum ether, extra pure, boiling range 100-140°C
CAS: 64742-49-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA233020025
Price (MYR)
536.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
536.00
BOT
Add to cart
Chemical Identifiers
CAS64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5
Specifications
Chemical Name or MaterialPetroleum ether
Melting Point-30.0°C
CAS64742-49-0
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Very toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.