Hexamethylenetetramine, 98.5%, contains an anticaking agent
CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
Have Questions?
Change view
Quantity:
2.5 kg
1 kg
Packaging:
Plastic bottle
Catalog number FSA232890025
Price (MYR)
725.00
BOT
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
725.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexamethylenetetramine
Name Note98.5%
CAS100-97-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement May cause an allergic skin reaction. Flammable solid.