Ethyl cellulose, ethoxyl content 48%, 300 cps
Ethyl cellulose, ethoxyl content 48%, 300 cps
Ethyl cellulose, ethoxyl content 48%, 300 cps
Ethyl cellulose, ethoxyl content 48%, 300 cps
Thermo Scientific Chemicals
Ethyl cellulose, ethoxyl content 48%, 300 cps
CAS: 9004-57-3 | C23H24N6O4 | 448.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA232705000
Price (MYR)
1,078.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,078.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl cellulose, ethoxyl
ColorWhite
CAS9004-57-3
Physical FormGranular Powder
Molecular FormulaC23H24N6O4