Linalyl acetate, 95%, synthetic
Linalyl acetate, 95%, synthetic
Linalyl acetate, 95%, synthetic
Linalyl acetate, 95%, synthetic
Thermo Scientific Chemicals
Linalyl acetate, 95%, synthetic
CAS: 115-95-7 | C12H20O2 | 196.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA232305000
Price (MYR)
642.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
642.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLinalyl acetate
Name Note95%
CAS115-95-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement