1-Hydroxypyrene, 99+%
1-Hydroxypyrene, 99+%
1-Hydroxypyrene, 99+%
1-Hydroxypyrene, 99+%
Thermo Scientific Chemicals
1-Hydroxypyrene, 99+%
CAS: 5315-79-7 | C16H10O | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
Packaging:
Glass bottle
Catalog number FSA232270500
Price (MYR)
538.00
EA
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
538.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hydroxypyrene
Name Note99+%
CAS5315-79-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning