Ethyl acetate, 99.5%, for analysis
Ethyl acetate, 99.5%, for analysis
Ethyl acetate, 99.5%, for analysis
Ethyl acetate, 99.5%, for analysis
Thermo Scientific Chemicals
Ethyl acetate, 99.5%, for analysis
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number FSA232110050
Price (MYR)
963.00
LT
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
963.00
LT
Add to cart
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-83.5°C
CAS141-78-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
View more
For analysis
RUO – Research Use Only