n-hexane, 95+%, for HPLC
n-hexane, 95+%, for HPLC
n-hexane, 95+%, for HPLC
n-hexane, 95+%, for HPLC
Thermo Scientific Chemicals
n-hexane, 95+%, for HPLC
CAS: 110-54-3 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA232100025
Price (MYR)
842.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
842.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Hexane
Melting Point-95.0°C
Name NoteFor HPLC
Absorbance(1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 250nm, 0.01 max. at 240nm, 0.03 max. at 230nm, 0.07 max. at 220nm, 0.25 max. at 210nm, 0.7 max. at 200nm
CAS110-54-3
View more
For HPLC
RUO – Research Use Only