1-Propanol, 99.5%, for analysis
1-Propanol, 99.5%, for analysis
1-Propanol, 99.5%, for analysis
1-Propanol, 99.5%, for analysis
Thermo Scientific Chemicals
1-Propanol, 99.5%, for analysis
CAS: 71-23-8 | C3H8O | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA232070025
Price (MYR)
1,015.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,015.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Propanol
Melting Point-127.0°C
CAS71-23-8
CAS Min %99.4
ColorBeige-Yellow to White
View more
For analysis
RUO – Research Use Only