Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Dodecyl sulfate, sodium salt, 99%, for biochemistry
Thermo Scientific Chemicals
Dodecyl sulfate, sodium salt, 99%, for biochemistry
CAS: 151-21-3 | C12H25NaO4S, C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Catalog number FSA230425000
Price (MYR)
1,726.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,726.00
BOT
Add to cart
Specifications
Assay99%
Chemical Name or MaterialDodecyl sulfate, sodium salt
CAS151-21-3
ColorWhite to Beige
Quantity500 g
View more
For biochemistry
RUO – Research Use Only