Rosolic acid
Rosolic acid
Rosolic acid
Thermo Scientific Chemicals
Rosolic acid
CAS: 603-45-2 | C19H14O3 | 290.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA229835000
Price (MYR)
6,434.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,434.00
EA
Add to cart
Specifications
Chemical Name or MaterialRosolic acid
CAS603-45-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
View more
Indicator
RUO – Research Use Only