Neutral Red, pure, high purity biological stain
Neutral Red, pure, high purity biological stain
Neutral Red, pure, high purity biological stain
Neutral Red, pure, high purity biological stain
Thermo Scientific Chemicals
Neutral Red, pure, high purity biological stain
CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA229810250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNeutral red
Melting Point290.0°C
Name NoteHigh purity biological stain
CAS553-24-2
Health Hazard 2GHS H Statement:
Suspected of causing genetic defects.
View more
Indicator
RUO – Research Use Only