Erioglaucine disodium salt, pure
Erioglaucine disodium salt, pure
Erioglaucine disodium salt, pure
Erioglaucine disodium salt, pure
Thermo Scientific Chemicals
Erioglaucine disodium salt, pure
CAS: 3844-45-9 | C37H34N2Na2O9S3 | 792.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA229730250
Price (MYR)
516.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
516.00
BOT
Add to cart
Specifications
Chemical Name or MaterialErioglaucine disodium salt
CAS3844-45-9
Absorbance1000 (at lambda min.)
Health Hazard 2GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.